accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 65753-93-7

IUPAC Name: 2-(dimethylamino)-N-hydroxy-N-methylacetamide
Synonyms: 2-(dimethylamino)-N-hydroxy-N-methylacetamide
Molecular Weight (g/mol): 132.16
Molecular Formula: C5H12N2O2
InChi Key: SEKVYKWEEZWQBZ-UHFFFAOYSA-N
SMILES: CN(C)CC(=O)N(C)O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results