Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl-2-dimethylaminoacetohydroxamic Acid 97.0+%, TCI America™
SDP

Catalog No. M13161G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13161G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M13161G Supplier TCI America Supplier No. M13161G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 65753-93-7
Molecular Formula C5H12N2O2
Molecular Weight (g/mol) 132.16
MDL Number MFCD01861290
InChI Key SEKVYKWEEZWQBZ-UHFFFAOYSA-N
Synonym 2-Dimethylamino-N-hydroxy-N-methylacetamide
PubChem CID 11040741
IUPAC Name 2-(dimethylamino)-N-hydroxy-N-methylacetamide
SMILES CN(C)CC(=O)N(C)O

Specifications

Melting Point 100°C
Color White
Quantity 1 g
Formula Weight 132.16
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Methyl-2-dimethylaminoacetohydroxamic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.