accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOOOFFF

CAS RN 658-78-6

IUPAC Name: 4-nitrophenyl 2,2,2-trifluoroacetate
Synonyms: 4-nitrophenyl 2,2,2-trifluoroacetate
Molecular Weight (g/mol): 235.12
Molecular Formula: C8H4F3NO4
InChi Key: JFOIBTLTZWOAIC-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(OC(=O)C(F)(F)F)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results