Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrophenyl Trifluoroacetate 98.0+%, TCI America™
SDP

Catalog No. T06815G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T06815G 5 g
T068125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T06815G Supplier TCI America Supplier No. T06815G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 658-78-6
Molecular Formula C8H4F3NO4
Molecular Weight (g/mol) 235.118
MDL Number MFCD00007324
InChI Key JFOIBTLTZWOAIC-UHFFFAOYSA-N
Synonym Trifluoroacetic Acid 4-Nitrophenyl Ester
PubChem CID 69569
IUPAC Name (4-nitrophenyl) 2,2,2-trifluoroacetate
SMILES C1=CC(=CC=C1[N+](=O)[O-])OC(=O)C(F)(F)F

Specifications

Melting Point 37°C
Color Yellow
Boiling Point 140°C
Quantity 5 g
Formula Weight 235.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Nitrophenyl Trifluoroacetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.