accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO

CAS RN 6581-66-4

IUPAC Name: 2-methoxyoxane
Synonyms: pyran, 2-methoxy-tetrahydro-
Molecular Weight (g/mol): 116.16
Molecular Formula: C6H12O2
InChi Key: XTDKZSUYCXHXJM-UHFFFAOYNA-N
SMILES: COC1CCCCO1
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results