accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 65855-02-9

IUPAC Name: (5R)-1-benzyl-5-ethoxyimidazolidine-2,4-dione
Synonyms: (5R)-1-benzyl-5-ethoxyimidazolidine-2,4-dione
Molecular Weight (g/mol): 234.26
Molecular Formula: C12H14N2O3
InChi Key: FUQZCDCFSMSNBP-LLVKDONJSA-N
SMILES: CCO[C@H]1N(CC2=CC=CC=C2)C(=O)NC1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results