Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzyl-5-ethoxyhydantoin 98.0+%, TCI America™
SDP

Catalog No. B308525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B308525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B308525G Supplier TCI America Supplier No. B308525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 65855-02-9
Molecular Formula C12H14N2O3
Molecular Weight (g/mol) 234.26
MDL Number MFCD03838588
InChI Key FUQZCDCFSMSNBP-LLVKDONJSA-N
PubChem CID 4079622
IUPAC Name (5R)-1-benzyl-5-ethoxyimidazolidine-2,4-dione
SMILES CCO[C@H]1N(CC2=CC=CC=C2)C(=O)NC1=O

Specifications

Melting Point 93°C
Color White-Yellow
Quantity 25 g
Formula Weight 234.26
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1-Benzyl-5-ethoxyhydantoin
EINECSNumber (5)-5255
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.