accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CN

CAS RN 65860-74-4

IUPAC Name: 4'-undecyl-[1,1'-biphenyl]-4-carbonitrile
Synonyms: 4'-undecyl-[1,1'-biphenyl]-4-carbonitrile
Molecular Weight (g/mol): 333.52
Molecular Formula: C24H31N
InChi Key: YIJBPYUXIFSTAP-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC1=CC=C(C=C1)C1=CC=C(C=C1)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results