accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiCH3H3COSiH3CCH3H3COSiCH3H3CCH3

CAS RN 65946-59-0

IUPAC Name: 2,2,6,6-tetramethyl-4-[(trimethylsilyl)methylidene]-3,5-dioxa-2,6-disilaheptane
Synonyms: 2,2,6,6-tetramethyl-4-[(trimethylsilyl)methylidene]-3,5-dioxa-2,6-disilaheptane
Molecular Weight (g/mol): 276.6
Molecular Formula: C11H28O2Si3
InChi Key: LIAZNEMTJSEKDC-UHFFFAOYSA-N
SMILES: C[Si](C)(C)OC(O[Si](C)(C)C)=C[Si](C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results