Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1-Bis(trimethylsilyloxy)-2-trimethylsilylethene, Thermo Scientific™

Catalog No. AC331015000
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC331015000 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC331015000 Supplier Thermo Scientific Chemicals Supplier No. 331015000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65946-59-0
Molecular Formula C11H28O2Si3
Molecular Weight (g/mol) 276.598
InChI Key LIAZNEMTJSEKDC-UHFFFAOYSA-N
Synonym 2,2,6,6-tetramethyl-4-trimethylsilyl methylidene-3,5-dioxa-2,6-disilaheptane, trimethylsilylketene di trimethylsilyl acetal, trimethylsilyl ketene bis trimethylsilyl acetal, 1,1-bis trimethylsilyloxy-2-trimethylsilylethene, trimethyl-2-trimethylsilyl-1-trimethylsilyloxyethenoxy silane, 3,5-dioxa-2,6-disilaheptane,2,2,6,6-tetramethyl-4-trimethylsilyl methylene
PubChem CID 2733565
IUPAC Name trimethyl-(2-trimethylsilyl-1-trimethylsilyloxyethenoxy)silane
SMILES C[Si](C)(C)C=C(O[Si](C)(C)C)O[Si](C)(C)C

Specifications

Quantity 500 mg
Formula Weight 276.6
Chemical Name or Material 1, 1-Bis(trimethylsilyloxy)-2-trimethylsilylethene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.