accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOClClOCH3

CAS RN 6597-78-0

IUPAC Name: methyl 3,6-dichloro-2-methoxybenzoate
Synonyms: methyl 3,6-dichloro-2-methoxybenzoate
Molecular Weight (g/mol): 235.06
Molecular Formula: C9H8Cl2O3
InChi Key: AWSBKDYHGOOSML-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(Cl)C=CC(Cl)=C1OC
Molecular Weight (g/mol) 
  • (2)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)