Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dicamba Methyl Ester Solution, 1 Analyte, 100μg/mL in Methanol, Ultra Scientific

Catalog No. USHB1611
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-HB-161-1 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-HB-161-1 Supplier Agilent Technologies Supplier No. HB1611
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6597-78-0 , 67-56-1
Molecular Formula C9H8Cl2O3
Molecular Weight (g/mol) 235.06
InChI Key AWSBKDYHGOOSML-UHFFFAOYSA-N
PubChem CID 81070
IUPAC Name methyl 3,6-dichloro-2-methoxybenzoate
SMILES COC1=C(C=CC(=C1C(=O)OC)Cl)Cl

Specifications

CAS Max % 99.98, 0.01
Physical Form Liquid
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 100μg/mL
Chemical Name or Material Dicamba Methyl Ester, 100μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.