accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONClNOO

CAS RN 65976-60-5

IUPAC Name: 4-(4-chloro-2-nitrophenyl)morpholine
Synonyms: 4-(4-chloro-2-nitrophenyl)morpholine
Molecular Weight (g/mol): 242.66
Molecular Formula: C10H11ClN2O3
InChi Key: KJISHWYSNGQNFM-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(C=CC(Cl)=C1)N1CCOCC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results