accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOO

CAS RN 66003-63-2

IUPAC Name: 12-ethoxy-12-oxododecanoate
Synonyms: 12-ethoxy-12-oxododecanoate
Molecular Weight (g/mol): 257.35
Molecular Formula: C14H25O4
InChi Key: VJOPNKODAZUCRH-UHFFFAOYSA-M
SMILES: CCOC(=O)CCCCCCCCCCC([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results