Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Monoethyl Dodecanedioate 98.0+%, TCI America™
SDP

Catalog No. D23745G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D23745G Supplier TCI America Supplier No. D23745G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 66003-63-2
Molecular Formula C14H25O4-
Molecular Weight (g/mol) 257.35
MDL Number MFCD00191643
InChI Key VJOPNKODAZUCRH-UHFFFAOYSA-M
Synonym Ethyl Hydrogen Dodecanedioate, Dodecanedioic Acid Monoethyl Ester
PubChem CID 20553525
IUPAC Name 12-ethoxy-12-oxododecanoate
SMILES CCOC(=O)CCCCCCCCCCC(=O)[O-]

Specifications

Melting Point 50°C
Color White
Boiling Point 184°C
Quantity 5 g
Formula Weight 258.36
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Monoethyl Dodecanedioate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.