CAS RN 66018-38-0
IUPAC Name:
8,9,16-trihydroxy-14-methoxy-3-methyl-3,4,7,8,9,10-hexahydro-1H-2-benzoxacyclotetradecine-1,7-dione
Synonyms:
8,9,16-trihydroxy-14-methoxy-3-methyl-4,8,9,10-tetrahydro-3H-2-benzoxacyclotetradecine-1,7-dione
Molecular Weight (g/mol):
362.38
Molecular Formula:
C19H22O7
InChi Key:
NEQZWEXWOFPKOT-UHFFFAOYNA-N
SMILES:
COC1=CC(O)=C2C(=C1)C=CCC(O)C(O)C(=O)C=CCC(C)OC2=O
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ (5Z)-7-Oxozeaenol, Curvularia sp.
(5Z)-7-Oxozeaenol, CAS 66018-38-0, is a cell-permeable, highly potent, selective, irreversible, and ATP-competitive inhibitor of MAPKKK TAK1 activity (IC50 = 8 nM).
2
5Z-7-Oxozeaenol
CAS: 66018-38-0 Molecular Formula: C19H22O7 Molecular Weight (g/mol): 362.378 MDL Number: MFCD12912700 InChI Key: NEQZWEXWOFPKOT-UHFFFAOYSA-N Synonym: 8,9,16-trihydroxy-14-methoxy-3-methyl-4,8,9,10-tetrahydro-3h-2-benzoxacyclotetradecine-1,7-dione PubChem CID: 53442201 IUPAC Name: 5,6,15-trihydroxy-17-methoxy-11-methyl-12-oxabicyclo[12.4.0]octadeca-1(14),2,8,15,17-pentaene-7,13-dione SMILES: CC1CC=CC(=O)C(C(CC=CC2=CC(=CC(=C2C(=O)O1)O)OC)O)O