accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6602-28-4

IUPAC Name: 3-hydroxy-1,2-dihydro-1λ²-pyridine-1,1-diium-2-id-1-olate
Synonyms: 3-hydroxy-1,2-dihydro-1λ²-pyridine-1,1-diium-2-id-1-olate
Molecular Weight (g/mol): 112.11
Molecular Formula: C5H6NO2
InChi Key: MXRXARQXORUYQO-UHFFFAOYNA-N
SMILES: OC1=CC=C[NH++]([O-])[CH-]1
Grade 
  • (2)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results