Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hydroxypyridine N-Oxide 96.0+%, TCI America™
SDP

Catalog No. H04925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04925G 5 g
H049225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H04925G Supplier TCI America Supplier No. H04925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 6602-28-4 | C5H6NO2 | 112.11 g/mol

Chemical Identifiers

CAS 6602-28-4
Molecular Formula C5H6NO2
Molecular Weight (g/mol) 112.11
MDL Number MFCD00006200
InChI Key MXRXARQXORUYQO-UHFFFAOYNA-N
Synonym 3-hydroxypyridine-n-oxide, 3-hydroxypyridine 1-oxide, 3-hydroxypyridine n-oxide, 3-pyridinol n-oxide, pyridin-3-ol 1-oxide, 3-pyridinol, 1-oxide, 3-hydroxypyridin-1-ium-1-olate, 3-pyridinol-1-oxide, 3-hydroxypyridine-1-oxide, 3-pyridinol 1-oxide
PubChem CID 23069
IUPAC Name 3-hydroxy-1,2-dihydro-1λ²-pyridine-1,1-diium-2-id-1-olate
SMILES OC1=CC=C[NH++]([O-])[CH-]1

Specifications

Melting Point 190°C
Quantity 5 g
Formula Weight 111.10
Percent Purity ≥96.0% (GC,T)
Grade Research
Physical Form Crystalline Powder
Chemical Name or Material 3-Hydroxypyridine N-Oxide
RTECSNumber UU7719100
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.