accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNBOH3CH3CH3CCH3O

CAS RN 660867-80-1

IUPAC Name: 2-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Synonyms: 2-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Weight (g/mol): 219.09
Molecular Formula: C12H18BNO2
InChi Key: MBTULFIFECUURA-UHFFFAOYSA-N
SMILES: CC1=CC(=CC=N1)B1OC(C)(C)C(C)(C)O1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results