accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3CH3H3CFBFFF

CAS RN 661-36-9

IUPAC Name: tetrafluoroboranuide; tetramethylazanium
Synonyms: tetramethylammonium ion tetrafluoroborate
Molecular Weight (g/mol): 160.95
Molecular Formula: C4H12BF4N
InChi Key: XWFABLFRYCHILB-UHFFFAOYSA-N
SMILES: C[N+](C)(C)C.F[B-](F)(F)F
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (1)
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)