Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethylammonium Tetrafluoroborate 98.0+%, TCI America™
SDP

Catalog No. T084225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T084225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T084225G Supplier TCI America Supplier No. T084225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 661-36-9
Molecular Formula C4H12BF4N
Molecular Weight (g/mol) 160.95
MDL Number MFCD00011745
InChI Key XWFABLFRYCHILB-UHFFFAOYSA-N
Synonym tetramethylammonium tetrafluoroborate, tetramethylammonium fluoroborate, borate 1-, tetrafluoro-, tetramethylammonium salt, tetramethylazanium tetrafluoroborate, n,n,n-trimethylmethanaminium tetrafluoroborate 1-, tetramethylammonium ion tetrafluoroborate, ammonium, tetramethyl-, tetrafluoroborate 1-, acmc-209ntt, methanaminium, n,n,n-trimethyl-, tetrafluoroborate 1-, tetramethyl ammonium tetrafluoroborate
PubChem CID 12621
IUPAC Name tetrafluoroboranuide; tetramethylazanium
SMILES C[N+](C)(C)C.F[B-](F)(F)F

Specifications

Melting Point 330°C
Color White
Quantity 25 g
UN Number 1759
Formula Weight 160.95
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Tetramethylammonium Tetrafluoroborate
EINECSNumber (2)-0186&(2)-3044
RTECSNumber BS8300000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.