accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6610-42-0

IUPAC Name: 2-(2-{[(benzyloxy)carbonyl]amino}-4-carbamoylbutanamido)acetic acid
Synonyms: (2-{[(benzyloxy)carbonyl]amino}-4-carbamoylbutanamido)acetic acid
Molecular Weight (g/mol): 337.33
Molecular Formula: C15H19N3O6
InChi Key: SOUXAAOTONMPRY-UHFFFAOYNA-N
SMILES: NC(=O)CCC(NC(=O)OCC1=CC=CC=C1)C(=O)NCC(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)