accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 66107-36-6

IUPAC Name: 2-chlorophenyl trifluoromethanesulfonate
Synonyms: 2-chlorophenyl trifluoromethanesulfonate
Molecular Weight (g/mol): 260.61
Molecular Formula: C7H4ClF3O3S
InChi Key: KTLNEJOQAOFUTO-UHFFFAOYSA-N
SMILES: FC(F)(F)S(=O)(=O)OC1=CC=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results