Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chlorophenyl Trifluoromethanesulfonate 97.0+%, TCI America™
SDP

Catalog No. C23265G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C23265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C23265G Supplier TCI America Supplier No. C23265G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 66107-36-6
Molecular Formula C7H4ClF3O3S
Molecular Weight (g/mol) 260.611
MDL Number MFCD07784322
InChI Key KTLNEJOQAOFUTO-UHFFFAOYSA-N
Synonym Trifluoromethanesulfonic Acid 2-Chlorophenyl Ester, 2-Chlorophenyl Triflate
PubChem CID 581421
IUPAC Name (2-chlorophenyl) trifluoromethanesulfonate
SMILES C1=CC=C(C(=C1)OS(=O)(=O)C(F)(F)F)Cl

Specifications

Color Yellow
Boiling Point 120°C
Quantity 5 g
Formula Weight 260.61
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chlorophenyl Trifluoromethanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.