accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 66108-95-0

IUPAC Name: N1,N3-bis(2,3-dihydroxypropyl)-5-[N-(2,3-dihydroxypropyl)acetamido]-2,4,6-triiodobenzene-1,3-dicarboxamide
Synonyms: iohexol exypaque iohexolum iohexol 350 omnipaque 70 iohexol preparation histodenz iohexol (substance) omnipaque nycodenz
Molecular Weight (g/mol): 821.14
Molecular Formula: C19H26I3N3O9
InChi Key: NTHXOOBQLCIOLC-UHFFFAOYNA-N
SMILES: CC(=O)N(CC(O)CO)C1=C(I)C(C(=O)NCC(O)CO)=C(I)C(C(=O)NCC(O)CO)=C1I
Molecular Weight (g/mol) 
  • (3)
  • (2)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results