accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 66131-78-0

IUPAC Name: N-(5-iodopyridin-2-yl)acetamide
Synonyms: N-(5-iodopyridin-2-yl)acetamide
Molecular Weight (g/mol): 262.05
Molecular Formula: C7H7IN2O
InChi Key: KUSIBMGCSKLUCE-UHFFFAOYSA-N
SMILES: CC(=O)NC1=NC=C(I)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results