Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetamido-5-iodopyridine 98.0+%, TCI America™
SDP

Catalog No. A30505G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A30505G 5 g
A30501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A30505G Supplier TCI America Supplier No. A30505G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 66131-78-0
Molecular Formula C7H7IN2O
Molecular Weight (g/mol) 262.05
MDL Number MFCD01315262
InChI Key KUSIBMGCSKLUCE-UHFFFAOYSA-N
PubChem CID 1488682
IUPAC Name N-(5-iodopyridin-2-yl)acetamide
SMILES CC(=O)NC1=NC=C(I)C=C1

Specifications

Melting Point 156°C
Color White-Yellow
Quantity 5 g
Formula Weight 262.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Acetamido-5-iodopyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.