accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNNNCl

CAS RN 66138-05-4

IUPAC Name: 5-ethyl-2,3-diphenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium chloride
Synonyms: 5-ethyl-2,3-diphenyl-1,2λ⁵,3,4-tetrazol-2-ylium chloride
Molecular Weight (g/mol): 286.76
Molecular Formula: C15H15ClN4
InChi Key: WDLRRKFNUOOGIW-UHFFFAOYSA-M
SMILES: [Cl-].CCC1=NN(C2=CC=CC=C2)[N+](=N1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results