accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrN

CAS RN 6621-59-6

IUPAC Name: 6-bromohexanenitrile
Synonyms: 6-bromohexanenitrile
Molecular Weight (g/mol): 176.06
Molecular Formula: C6H10BrN
InChi Key: PHOSWLARCIBBJZ-UHFFFAOYSA-N
SMILES: BrCCCCCC#N
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)