Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Bromocapronitrile, MP Biomedicals

Catalog No. ICN214350 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN214350 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN214350 Supplier MP Biomedicals, Inc Supplier No. 0521435005

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6621-59-6
Molecular Formula C6H10BrN
Molecular Weight (g/mol) 176.06
MDL Number MFCD00013834
InChI Key PHOSWLARCIBBJZ-UHFFFAOYSA-N
Synonym 6-bromocapronitrile, 6-bromohexanonitrile, hexanenitrile, 6-bromo, 5-bromopentyl cyanide, 6-bromo-hexanenitrile, 6-bromanylhexanenitrile, pubchem12505, acmc-1b5au, 6-bromohexanenitrile, phoswlarcibbjz-uhfffaoysa
PubChem CID 81093
IUPAC Name 6-bromohexanenitrile
SMILES BrCCCCCC#N

Specifications

Quantity 5 g
Formula Weight 176.06
Chemical Name or Material 6-Bromocapronitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.