accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 66232-57-3

IUPAC Name: 2-methyl-6-nitrobenzoyl chloride
Synonyms: 2-methyl-6-nitrobenzoyl chloride
Molecular Weight (g/mol): 199.59
Molecular Formula: C8H6ClNO3
InChi Key: ZMAJSTDJAIVHGZ-UHFFFAOYSA-N
SMILES: CC1=C(C(Cl)=O)C(=CC=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results