accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOON

CAS RN 6624-49-3

IUPAC Name: isoquinoline-3-carboxylic acid
Synonyms: isoquinoline-3-carboxylic acid
Molecular Weight (g/mol): 173.17
Molecular Formula: C10H7NO2
InChi Key: KVMMIDQDXZOPAB-UHFFFAOYSA-N
SMILES: OC(=O)C1=NC=C2C=CC=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results