Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Isoquinoline-3-carboxylic acid, 97%, Thermo Scientific™

Catalog No. AC454490010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC454490010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC454490010 Supplier Thermo Scientific Chemicals Supplier No. 454490010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

Chemical Identifiers

CAS 6624-49-3
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.17
InChI Key KVMMIDQDXZOPAB-UHFFFAOYSA-N
Synonym 3-isoquinolinecarboxylic acid, 3-carboxyisoquinoline, 3-isoquinoline carboxylic acid, 3-isoquinaldic acid, pubchem16889, maybridge1_002411, acmc-1b7v6, isoquinoline-3-caboxylic acid, ksc217c5f, isoquinoline 3-carboxylic acid
PubChem CID 124656
IUPAC Name isoquinoline-3-carboxylic acid
SMILES C1=CC=C2C=NC(=CC2=C1)C(=O)O

Specifications

Melting Point -170°C to 171.5°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 173.17
Percent Purity 97%
Chemical Name or Material Isoquinoline-3-carboxylic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. Harmful if inhaled. May cause respiratory irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.