accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3Br

CAS RN 6627-78-7

IUPAC Name: 1-bromo-4-methylnaphthalene
Synonyms: 1-bromo-4-methylnaphthalene
Molecular Weight (g/mol): 221.1
Molecular Formula: C11H9Br
InChi Key: IDRVLLRKAAHOBP-UHFFFAOYSA-N
SMILES: CC1=C2C=CC=CC2=C(Br)C=C1
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results