accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 66356-53-4

IUPAC Name: 2-[({5-[(dimethylamino)methyl]furan-2-yl}methyl)sulfanyl]ethan-1-amine
Synonyms: 2-[({5-[(dimethylamino)methyl]furan-2-yl}methyl)sulfanyl]ethanamine
Molecular Weight (g/mol): 214.33
Molecular Formula: C10H18N2OS
InChi Key: JFGCGQJHMUYGLU-UHFFFAOYSA-N
SMILES: CN(C)CC1=CC=C(CSCCN)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results