Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-[((5-[(dimethylamino)methyl]-2-furyl)methyl)thio]ethan-1-amine, Thermo Scientific™

Catalog No. SB01748DE
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SB01748DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SB01748DE Supplier Thermo Scientific Chemicals Supplier No. SB01748DE
Add to Cart

Chemical Identifiers

CAS 66356-53-4
Molecular Formula C10H18N2OS
Molecular Weight (g/mol) 214.327
InChI Key JFGCGQJHMUYGLU-UHFFFAOYSA-N
Synonym 2-2-aminoethylthio methyl-5-n,n-dimethylamino methyl furan, unii-s8w8i89w43, 2-5-dimethylamino methylfuryl thio ethylamine, 2-2-aminoethylthiomethyl-5-dimethylaminomethylfuran, 5-2-aminoethyl thiomethyl-n,n-dimethyl-2-furanmethanamine, 2-furanmethanamine, 5-2-aminoethyl thio methyl-n,n-dimethyl, 2-5-dimethylamino methyl-2-furyl methylsulfanyl ethanamine, 5-2-aminoethyl thio methyl-n,n-dimethyl-2-furanmethanamine, 2-5-dimethylamino methyl-2-furyl methyl thio ethan-1-amine
PubChem CID 162203
IUPAC Name 2-[[5-[(dimethylamino)methyl]furan-2-yl]methylsulfanyl]ethanamine
SMILES CN(C)CC1=CC=C(O1)CSCCN

Specifications

Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 5 g
Formula Weight 214.33
Percent Purity 95%
Chemical Name or Material 2-[({5-[(Dimethylamino)methyl]-2-furyl}methyl)thio]ethan-1-amine
Hazard Category GHS Signal Word: Danger
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.