accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6639-30-1

CH3ClClCl

CAS RN 6639-30-1

IUPAC Name: 1,2,4-trichloro-5-methylbenzene
Synonyms: 2,4,5-trichlorotoluene
Molecular Weight (g/mol): 195.47
Molecular Formula: C7H5Cl3
InChi Key: ZCXHZKNWIYVQNC-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=C(Cl)C=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results