accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6640-23-9

IUPAC Name: pyrimidine-4,5,6-triamine sulfuric acid hydrate
Synonyms: pyrimidine-4,5,6-triamine sulfuric acid hydrate
Molecular Weight (g/mol): 241.22
Molecular Formula: C4H11N5O5S
InChi Key: WRXLIMZODSSQIH-UHFFFAOYSA-N
SMILES: O.OS(O)(=O)=O.NC1=NC=NC(N)=C1N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results