Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5,6-Triaminopyrimidine Sulfate Hydrate 98.0+%, TCI America™
SDP

Catalog No. T03361G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T03361G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T03361G Supplier TCI America Supplier No. T03361G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6640-23-9
Molecular Formula C4H11N5O5S
Molecular Weight (g/mol) 241.222
MDL Number MFCD00012789
InChI Key WRXLIMZODSSQIH-UHFFFAOYSA-N
Synonym 4,5,6-triaminopyrimidine sulfate hydrate, 4,5,6-triaminepyrimidine sulfate hydrate, pyrimidine, 4,5,6-triamino-, sulfate, hydrate, pyrimidine-4,5,6-triamine sulfate hydrate, pyrimidine-4,5,6-triamine sulfuric acid hydrate, pyrimidine-4,5,6-triamine; sulfuric acid; hydrate, zlchem 873, acmc-209nvb, c4h7n5.h2o.h2so4, 4,5,6-triaminopyrimidine sulfate, hydrate
PubChem CID 201714
IUPAC Name pyrimidine-4,5,6-triamine;sulfuric acid;hydrate
SMILES C1=NC(=C(C(=N1)N)N)N.O.OS(=O)(=O)O

Specifications

Quantity 1 g
Formula Weight 223.21
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4,5,6-Triaminopyrimidine Sulfate Hydrate
RTECSNumber UW5825000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.