accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 66417-30-9

IUPAC Name: 2-bromo-1-methyl-4-(trifluoromethyl)benzene
Synonyms: 2-bromo-1-methyl-4-(trifluoromethyl)benzene
Molecular Weight (g/mol): 239.04
Molecular Formula: C8H6BrF3
InChi Key: TWWUWLMGYSLODU-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1Br)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results