accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3(S)NH2ONHOOCH3

CAS RN 66447-31-2

IUPAC Name: (2S)-2-amino-4-methyl-N-(4-methyl-2-oxo-2H-chromen-7-yl)pentanamide
Synonyms: (2S)-2-amino-4-methyl-N-(4-methyl-2-oxochromen-7-yl)pentanamide
Molecular Weight (g/mol): 288.35
Molecular Formula: C16H20N2O3
InChi Key: GTAAIHRZANUVJS-ZDUSSCGKSA-N
SMILES: CC(C)C[C@H](N)C(=O)NC1=CC=C2C(C)=CC(=O)OC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)