Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals L-Leucine 7-amido-4-methylcoumarin hydrate

Catalog No. AAJ64951MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64951MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ64951MD Supplier Thermo Scientific Chemicals Supplier No. J64951MD
Add to Cart

CAS: 66447-31-2 | C16H20N2O3 | 288.35 g/mol

A sensitive fluorogenic substrate for leucine aminopeptidase and aminopeptidase M

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66447-31-2
Molecular Formula C16H20N2O3
Molecular Weight (g/mol) 288.35
MDL Number MFCD04966490
InChI Key GTAAIHRZANUVJS-ZDUSSCGKSA-N
Synonym h-leu-amc, 7-leucylamido-4-methylcoumarin, 2s-2-amino-4-methyl-n-4-methyl-2-oxochromen-7-yl pentanamide, l-leucine 7-amido-4-methylcoumarin hydrate, s-2-amino-4-methyl-n-4-methyl-2-oxo-2h-chromen-7-yl pentanamide, l-leu-amc, l-l-amc, leu-mca, ambotzhaa1181, h-leu-mca free base
PubChem CID 171787
IUPAC Name (2S)-2-amino-4-methyl-N-(4-methyl-2-oxochromen-7-yl)pentanamide
SMILES CC(C)C[C@H](N)C(=O)NC1=CC=C2C(C)=CC(=O)OC2=C1

Specifications

Color White to Yellow
Quantity 250 mg
Formula Weight 288.37 (Anhydrous)
Physical Form Powder
Chemical Name or Material L-Leucine 7-amido-4-methylcoumarin hydrate
EINECSNumber 266-363-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.