accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6645-46-1

IUPAC Name: [(2R)-3-carboxy-2-hydroxypropyl]trimethylazanium chloride
Synonyms: carnitine chloride|levocarnitine hydrochloride
Molecular Weight (g/mol): 197.66
Molecular Formula: C7H16ClNO3
InChi Key: JXXCENBLGFBQJM-FYZOBXCZSA-N
SMILES: [Cl-].C[N+](C)(C)C[C@H](O)CC(O)=O
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (3)
  • (3)
Physical Form 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (5)