Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Carnitine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. C30585G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30585G 5 g
C305825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C30585G Supplier TCI America Supplier No. C30585G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6645-46-1
Molecular Formula C7H16ClNO3
Molecular Weight (g/mol) 197.66
MDL Number MFCD00066100
InChI Key JXXCENBLGFBQJM-FYZOBXCZSA-N
Synonym r-3-carboxy-2-hydroxy-n,n,n-trimethylpropan-1-aminium chloride, l-carnitine hcl, unii-j3y5e6iks3, l-carnitine hydrochloride, levocarnitine chloride, j3y5e6iks3, r-carnitine hydrochloride, carnitine l-form hydrochloride, l-3-carboxy-2-hydroxypropyl trimethylammonium chloride
PubChem CID 656657
IUPAC Name [(2R)-3-carboxy-2-hydroxypropyl]trimethylazanium chloride
SMILES [Cl-].C[N+](C)(C)C[C@H](O)CC(O)=O

Specifications

Melting Point 139°C
Color White
Quantity 5 g
Formula Weight 197.66
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Carnitine Hydrochloride
EINECSNumber (9)-0253
RTECSNumber BP2979100
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.