accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 66472-86-4

IUPAC Name: bis((3-aminophenyl)boronic acid); sulfuric acid
Synonyms: bis(M-aminophenylboronic acid); sulfuric acid
Molecular Weight (g/mol): 371.96
Molecular Formula: C12H18B2N2O8S
InChi Key: UKTAURVTSWDIQR-UHFFFAOYSA-N
SMILES: OS(O)(=O)=O.NC1=CC=CC(=C1)B(O)O.NC1=CC=CC(=C1)B(O)O
Molecular Weight (g/mol) 
  • (2)
  • (8)
  • (2)
Quantity 
  • (3)
  • (5)
  • (4)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (5)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (4)
  • (3)
  • (5)
  • (2)
  • (8)
  • (2)
  • (3)
  • (5)
  • (4)
  • (3)
  • (2)
  • (5)
  • (2)