Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3-Aminophenylboronic Acid Hemisulfate Salt

Catalog No. A717515G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A717515G 5 g
A7175125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A717515G Supplier Sigma Aldrich Supplier No. A717515G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3-Aminophenylboronic Acid Hemisulfate Salt

Specifications

CAS 66472-86-4
Density 0.965 g/mL at 25°C
Boiling Point 72°C to 73°C (11 mmHg, lit.)
Molecular Formula C3H9NO
Linear Formula CH3CH(NH2)CH2OH
Quantity 5 g
MDL Number MFCD00064412
Synonym (3-Aminophenyl)boric acid hemisulfate; (3-aminophenyl)-boronic acid sulfate (2:1); (m-Aminophenyl)boronic acid hemisulfate; 3-Aminobenzeneboronic acid hemisulfate salt; Bis[3-dihydroxyboranyl)benzenaminium] sulfate
Molecular Weight (g/mol) 75.11 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.