accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 664985-43-7

IUPAC Name: 7-chloro-N-[2-(dimethylamino)ethyl]-2,1,3-benzoxadiazole-4-sulfonamide
Synonyms: 7-chloro-N-[2-(dimethylamino)ethyl]-2,1,3-benzoxadiazole-4-sulfonamide
Molecular Weight (g/mol): 304.75
Molecular Formula: C10H13ClN4O3S
InChi Key: RJEYFWVHUXSWTA-UHFFFAOYSA-N
SMILES: CN(C)CCNS(=O)(=O)C1=CC=C(Cl)C2=NON=C12
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results