Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DAABD-Cl [=4-[2-(Dimethylamino)ethylaminosulfonyl]-7-chloro-2,1,3-benzoxadiazole], TCI America™
SDP

Catalog No. A5596100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A5596100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A5596100MG Supplier TCI America Supplier No. A5596100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

[for Proteome Analysis]

Chemical Identifiers

CAS 664985-43-7
Molecular Formula C10H13ClN4O3S
Molecular Weight (g/mol) 304.749
InChI Key RJEYFWVHUXSWTA-UHFFFAOYSA-N
Synonym 4-[2-(Dimethylamino)ethylaminosulfonyl]-7-chloro-2,1,3-benzoxadiazole
PubChem CID 44629802
IUPAC Name 4-chloro-N-[2-(dimethylamino)ethyl]-2,1,3-benzoxadiazole-7-sulfonamide
SMILES CN(C)CCNS(=O)(=O)C1=CC=C(C2=NON=C12)Cl

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 304.75
Physical Form Crystalline Powder
Chemical Name or Material DAABD-Cl [=4-[2-(Dimethylamino)ethylaminosulfonyl]-7-chloro-2,1,3-benzoxadiazole] [for Proteo
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.