accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 665-49-6

IUPAC Name: tetraethylphosphanium; tetrafluoroboranuide
Synonyms: tetraethylphosphanium tetrafluoroborate
Molecular Weight (g/mol): 234.03
Molecular Formula: C8H20BF4P
InChi Key: IOQILLMTUOVXHI-UHFFFAOYSA-N
SMILES: F[B-](F)(F)F.CC[P+](CC)(CC)CC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results