Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraethylphosphonium Tetrafluoroborate 97.0+%, TCI America™
SDP

Catalog No. T17475G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T17475G 5 g
T174725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T17475G Supplier TCI America Supplier No. T17475G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 665-49-6
Molecular Formula C8H20BF4P
Molecular Weight (g/mol) 234.025
MDL Number MFCD01631314
InChI Key IOQILLMTUOVXHI-UHFFFAOYSA-N
PubChem CID 16218631
IUPAC Name tetraethylphosphanium;tetrafluoroborate
SMILES [B-](F)(F)(F)F.CC[P+](CC)(CC)CC

Specifications

Color White
Quantity 5 g
UN Number 1759
Formula Weight 234.03
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Tetraethylphosphonium Tetrafluoroborate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.